missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ T-CAL™ Turbidity Standard, 10 NTU

Catalog No. 15398366
Change view
Click to view available options
Volume (Metric):
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
15398366 125 mL
15398367 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15398366 Supplier Lovibond™ Supplier No. 48011212

T-CAL™ Turbidity Standards are stabilized Formazin standards prepared in ready-to-use concentrations to ensure accurate turbidity measurements. Eliminate the opportunity for inaccurate turbidimeter calibrations resulting from the improper dilution or handling of Formazin stock solution.

  • Save time and eliminate errors - no need for dilution
  • EPA approved primary turbidity standard
  • Long shelf life
  • All concentrations of T-CAL™ Turbidity Standards are verified to meet tight quality control specifications, are EPA and ISO compliant for reporting purposes and guaranteed to save you time. As a primary standard, T-CAL™ Turbidity Standards can be used to calibrate or verify any manufacturer's turbidimeter.

Specifications

Type Primary Turbidity Standard
For Use With (Equipment) Turbidimeter
Calibration Type Primary
Form Liquid
For Use With (Application) Calibration or Verification of a Turbidimeter
Turbidity 10 NTU
Certifications/Compliance EN ISO 7027, USEPA 180.1 and APHA 2130 Part B.
Volume (Metric) 125 mL
Quantity 1 Ea.
Packaging Type Amber Plastic Bottles