Learn More
Can-Am Instruments Ltd T-BAR SPINDLE T-B
Catalog No. NC9446630
Encompass
This item has a minimum order quantity of 2 per supplier requirements. T-Bar Spindle, Entry Code: 92, SMC: 40, SRC: 0, Entry Code Allows DV-II+Pro to Calculate Viscosity, Shear Rate and Shear Stress Values, SMC for Viscosity and Shear Stress Calculations, SRC Used for Shear Rate and Shear Stress Calculations
Catalog No. NC9446630 Supplier Can-Am Instruments Ltd Supplier No. TB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This item has a minimum order quantity of 2 per supplier requirements. T-Bar Spindle, Entry Code: 92, SMC: 40, SRC: 0, Entry Code Allows DV-II+Pro to Calculate Viscosity, Shear Rate and Shear Stress Values, SMC for Viscosity and Shear Stress Calculations, SRC Used for Shear Rate and Shear Stress Calculations