missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T-1270 Fixed Angle Titanium Rotor

Catalog No. 08259
Change view
Click to view available options
Capacity:
12 x 12.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
08259 12 x 12.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08259 Supplier Thermo Scientific™ Supplier No. 08259

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Effectively perform a wide range of applications with the T-1270 Rotor, which offers 12-place capacity.

Effectively perform a wide range of applications with the Thermo Scientific™ T-1270 Fixed Angle Rotor, which offers 12-place capacity.

Recommended for:

DNA isolation, serum flotation.

Specifications

Description T-1270 Fixed Angle Titanium Rotor
Material Titanium
Type Fixed Angle Rotor
Capacity 12 x 12.5 mL
Max. RCF 448,811 x g
Max. Speed 70,000 rpm
Radius (Metric) 82 mm
Radius (Metric) Min. 40.6 mm
K Factor 36.3
Max. Tube Length 80 mm
Max. Tube Diameter 16 mm
Net Weight 13 lb. (5.9 kg)
For Use With (Equipment) Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle 24°
Avg. RCF 335,513 x g
Min. RCF 222,216 x g
Show More Show Less