missing translation for 'onlineSavingsMsg'
Learn More

Syringic Acid 97.0+%, TCI America™

Catalog No. G0014500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0014500G 500 g
G001425G 25 g
G0014100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0014500G Supplier TCI America Supplier No. G0014500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 530-57-4
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.17
MDL Number MFCD00002552
InChI Key JMSVCTWVEWCHDZ-UHFFFAOYSA-N
Synonym syringic acid, 3,5-dimethoxy-4-hydroxybenzoic acid, cedar acid, gallic acid 3,5-dimethyl ether, benzoic acid, 4-hydroxy-3,5-dimethoxy, 3,5-dimethoxy-4-hydroxybenzyl acid, 4-hydroxy-3,5-dimethoxy-benzoic acid, chembl1414, syringicacid, syringlicacid
PubChem CID 10742
ChEBI CHEBI:68329
IUPAC Name 4-hydroxy-3,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1O)C(O)=O

Specifications

Melting Point 209°C
Quantity 500 g
Formula Weight 198.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Syringic Acid
EINECSNumber (3)-3963
RTECSNumber DH2090000
TSCA Yes