Learn More
Chemglass Life Sciences Syringe, Luer Lock, 2cc Capacity, 1/10cc Subdivisions
Supplier: Chemglass Inc CG307001
Syringe, Luer Lock, 2cc Capacity, 1/10cc Subdivisions. Borosilicate glass syringe has interchangeable barrels and plungers. Colored plunger ends identify the capacity of the syringe. Flanges are flat on two sides to prevent syringe from rolling. All sizes of syringes are equipped with a metal luer lock and permanently fused graduations. For needles see CG-3075.