Learn More
Chemglass Inc Syringe, Micro, Gas-Tight, 25mL capacity, PTFE Luer Lock
Supplier: Chemglass Inc CG307436
Syringe, Micro, Gas-Tight, 25mL capacity, PTFE Luer Lock. Syringes are designed for use in both liquid and gas chromatography. Due to the all-PTFE tip, they can also handle highly corrosive or Ultra-pure liquids. With the exacting tolerances held in the manufacture of these syringes, all components are interchangeable within each size range.PLEASE NOTE: PTFE LUER TIP OR LUER LOCK SYRINGES ARE NOT SUPPLIED WITH NEEDLES. FOR NEEDLES SEE CG-3075.