missing translation for 'onlineSavingsMsg'
Learn More

Syringaldehyde 98.0+%, TCI America™

Catalog No. p-7126336
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06351G 1 g
D063525G 25 g
D0635100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D06351G Supplier TCI America Supplier No. D06351G
Only null left

Chemical Identifiers

CAS 134-96-3
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00006943
InChI Key KCDXJAYRVLXPFO-UHFFFAOYSA-N
Synonym syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
PubChem CID 8655
ChEBI CHEBI:67380
IUPAC Name 4-hydroxy-3,5-dimethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(OC)=C1O

Specifications

Melting Point 112°C
Boiling Point 193°C
Quantity 1 g
Formula Weight 182.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Syringaldehyde
EINECSNumber (3)-1207
RTECSNumber CU5760000
TSCA Yes