missing translation for 'onlineSavingsMsg'
Learn More

Syringaldehyde, 98+%

Catalog No. AAA1293322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1293322 100 g
AAA1293306 5 g
AAA1293314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1293322 Supplier Thermo Scientific Chemicals Supplier No. A1293322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134-96-3 | C9H10O4 | 182.18 g/mol

Syringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Syringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.

Solubility
Very sparingly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to air. Incompatible with strong oxidizing agents, and bases.

Chemical Identifiers

CAS 134-96-3
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00006943
InChI Key KCDXJAYRVLXPFO-UHFFFAOYSA-N
Synonym syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
PubChem CID 8655
ChEBI CHEBI:67380
IUPAC Name 4-hydroxy-3,5-dimethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(OC)=C1O

Specifications

Melting Point 110°C to 113°C
Boiling Point 192°C to 193°C (14 mmHg)
Quantity 100 g
Beilstein 784514
Sensitivity Air Sensitive
Merck Index 14,9015
Solubility Information Very sparingly soluble in water.
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material Syringaldehyde
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-167-5
RTECSNumber CU5760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only