Learn More
Synthware™ Vacuum Transfer Adapter with PTFE Stopcocks
Adapter with 2mm stopcock used for the transfer of sensitive and hazardous solutions
$390.93 - $400.00
Specifications
| For Use With (Application) | Organic Synthesis |
|---|---|
| Material | Borosilicate Glass (3.3) |
| Product Type | Glass Joint Adapter |
| Quantity | 1 |
Description
Synthware vacuum transfer adapter with PTFE stopcocks and standard taper inner joint. Used for the transfer of sensitive and hazardous solutions while under vacuum.Specifications
| Organic Synthesis | |
| Borosilicate Glass (3.3) | |
| Glass Joint Adapter | |
| 1 |