Learn More
Synthware™ Vacuum or Gas Adapters
With hose connection on the top and male joint
$46.57 - $56.61
Specifications
Material Borosilicate Glass (3.3)
Product Type Vacuum or Gas Adapter
View More Specs
Products 4
Catalog Number Mfr. No. Standard Taper Joint No. Price Quantity  
Catalog Number Mfr. No. Standard Taper Joint No. Price Quantity  
31500536
View Documents
Synthware™
A301420
14/20 N/A
Only null left
31500537
View Documents
Synthware™
A301922
19/22 N/A
Only null left
31500538
View Documents
Synthware™
A302440
24/40 N/A
Only null left
31500539
View Documents
Synthware™
A302942
29/42 N/A
Only null left
Description
Description
Synthware vacuum or argon/nitrogen inlet adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure.
Specifications
Specifications
Borosilicate Glass (3.3)
Vacuum or Gas Adapter
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'