Learn More
Synthware™ Vacuum Adapter, with PTFE Stopcock, Microscale
Vacuum adapter with 1:5 PTFE stopcock. For microscale applications
Supplier: Synthware™ A381410
Description
- With 1:5 PTFE stopcock
- Joint 14/10
- Hose connection 8mm O.D.
Specifications
| Hose connection: 8mm O.D. | |
| 14/10 |