Learn More
Synthware™ Sure-Grip Separatory Funnels
GREENER_CHOICE
With 4 mm bore PTFE stopcock and No. 27 flask length glass or PTFE stopper
$357.50 - $635.64
Specifications
Material Glass
Stopper Number 27
Product Type Separatory funnel, Sure-grip
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Stopper Material Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Stopper Material Price Quantity & Availability  
31501687
GREENER_CHOICE
View Documents Promotion Details
Synthware™
F687500A
500 mL Glass
Each for $357.50
Only null left
31501688
GREENER_CHOICE
View Documents Promotion Details
Synthware™
F687000A
1000 mL Glass
Each for $476.86
Only null left
31501689
GREENER_CHOICE
View Documents Promotion Details
Synthware™
F687000
1000 mL PTFE
Each for $502.11
Only null left
31501690
GREENER_CHOICE
View Documents Promotion Details
Synthware™
F68272LA
2000 mL Glass
Each for $598.18
Only null left
31501691
GREENER_CHOICE
View Documents Promotion Details
Synthware™
F68272L
2000 mL PTFE
Each for $635.64
Only null left
Description
Description
  • With long stopper neck and straight section between funnel body and stopcock
  • Stopper: Glass and PFTE
Specifications
Specifications
Glass
27
Separatory funnel, Sure-grip