Learn More
Synthware™ Storage Flasks with Side Arm
Supplied with high vacuum valve with PTFE O-ring at tip
Supplier: Synthware™ F464508
Description
SYNTHWARE storage flask with standard taper outer joint and high vacuum valve. Valve has a PTFE O-ring at the tip. Valve is used to control liquid or gas flow into the flask.Specifications
| Clear | |
| 500 mL | |
| Organic Synthesis |
| Valve Size: 0-8mm | |
| Borosilicate Glass (3.3) | |
| 24/40 |