Learn More
Synthware™ Squibb Separatory Funnels with Glass Stopcock
GREENER_CHOICE
Supplied complete with flask length glass stopper
Supplier:  Synthware™ F780500
 View more versions of this product
Catalog No. 31501685
Only null left
Description
Description
Synthware Squibb Separatory Funnel is used to separate two immiscible liquids of different densities.
Specifications
Chemical Identifiers
Glass
Organic Synthesis
Glass
Clear
1
Squibb Separatory Funnel
Specifications
500 mL
Clear
1
Glass
Squibb Separatory Funnel
Glass
Organic Synthesis
4mm
27
Product Suggestions
Product Suggestions