Learn More
Synthware™ Squibb Separatory Funnels with Glass Stopcock
GREENER_CHOICE
Supplied complete with flask length glass stopper
Supplier:  Synthware™ F780125
 View more versions of this product
Catalog No. 31501683
Only null left
Description
Description
Synthware Squibb Separatory Funnel is used to separate two immiscible liquids of different densities.
Specifications
Chemical Identifiers
Glass
Organic Synthesis
Glass
Clear
1
Squibb Separatory Funnel
Specifications
125 mL
Clear
1
Glass
Squibb Separatory Funnel
Glass
Organic Synthesis
2mm
22
Product Suggestions
Product Suggestions