Learn More
Synthware™ Solvent Transfer/Storage Flasks
For long term deoxygenated solvent storage
Supplier: Synthware™ F481500
Description
- Center neck joint sealed off from direct access to flask
- A syringe or cannula is used to recover the solvent through valve
Specifications
| Valve Size: 0-4mm | |
| 500 mL | |
| 14/20 |