Learn More
Synthware™ Separatory Funnel, with PTFE Stopper
GREENER_CHOICE
Separatory Funnel includes PTFE Stopper.
Supplier:  Synthware™ F479250
 View more versions of this product
Catalog No. 31501651
Only null left
Description
Description
  • With upper outer standard taper joint.
Specifications
Chemical Identifiers
Borosilicate Glass (3.3)
PTFE
Separatory Funnel
Specifications
250 mL
19/22
PTFE
Borosilicate Glass (3.3)
4 mm
Separatory Funnel
Product Suggestions
Product Suggestions