Learn More
Synthware™ Separatory Funnel, with PTFE Stopper
Separatory Funnel includes PTFE Stopper.
Supplier: Synthware™ F474125
Description
- With upper outer standard taper joint.
Chemical Identifiers
| Borosilicate Glass (3.3) | |
| PTFE | |
| Separatory Funnel |
Specifications
| 125 mL | |
| 24/40 | |
| PTFE |
| Borosilicate Glass (3.3) | |
| 2 mm | |
| Separatory Funnel |