Learn More
Synthware™ Separatory Funnels with PTFE Stopcock and Stopper
Includes PTFE Stopper
Supplier: Synthware™ F459125
Description
- Upper outer and lower inner standard taper joints are same size
Specifications
| 125 mL | |
| 19/22 | |
| PTFE |
| Glass | |
| 2mm | |
| Separatory Funnel |