Learn More
Synthware™ Separatory Funnels with PTFE Stopcock and Stopper
Includes PTFE Stopper
Supplier:  Synthware™ F451125
 View more versions of this product
Catalog No. 31501621
Only null left
Explore available promotions
Description
Description
  • Upper outer and lower inner standard taper joints are same size
Specifications
Specifications
125 mL
14/20
PTFE
Glass
2mm
Separatory Funnel
Product Suggestions
Product Suggestions