missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Rotary Evaporator Anti-Splash Adapters, Modified, with Fritted Disc

Used when foam is likely to be created and drawn into the condenser of rotary evaporator
Supplier: Synthware™ A461250
Description
- With two 4mm drain holes at base of inner tube to allow solvent to drain back into evaporation flask
- Extra coarse fritted disc sealed above lower joint reduces foam overflow from being drawn into condenser
Chemical Identifiers
| Borosilicate Glass (3.3) | |
| Anti-splash Adapter with Fritted Disc |
Specifications
| Borosilicate Glass (3.3) | |
| 250 mL | |
| Anti-splash Adapter with Fritted Disc | |
| Upper: 24/40, Lower: 14/20 |