Learn More
Synthware™ Reflux Condenser, with Removable Hose Connections
GREENER_CHOICE
For use with volatile solvents
Supplier:  Synthware™ C564125
 View more versions of this product
Catalog No. 31500750
Only null left
Description
Description
  • Coil style reflux condenser with tightly wrapped coil providing maximum cooling
  • With top outer standard taper joint and lower inner drip tip joint
Specifications
Chemical Identifiers
Glass
Reflux Condenser
Specifications
24/40
285 mm
Hose connection: 10mm O.D.
Glass
125 mm
Reflux Condenser
Product Suggestions
Product Suggestions