Learn More
Synthware™ Reflux Condenser, with Removable Hose Connections
GREENER_CHOICE
For use with volatile solvents
Supplier:  Synthware™ C562280
 View more versions of this product
Catalog No. 31500749
Only null left
Description
Description
  • Coil style reflux condenser with tightly wrapped coil providing maximum cooling
  • With top outer standard taper joint and lower inner drip tip joint
Specifications
Chemical Identifiers
Glass
Reflux Condenser
Specifications
14/20
Hose connection: 8mm O.D.
305 mm
Glass
180 mm
Reflux Condenser
Product Suggestions
Product Suggestions