Learn More
Synthware™ Reducing Adapters
With standard taper joints at top and bottom
Supplier: Synthware™ A202434
Description
Synthware reducing adapters are used for connecting dissimilar standard taper joint sizes. The lower inner joint is larger in size than the top outer joint.Specifications
| Organic Synthesis | |
| Reducing adapter | |
| Top: 24/40, Bottom: 34/45 |
| Borosilicate Glass (3.3) | |
| 1 |