Learn More
Synthware™ Recovery Flasks
GREENER_CHOICE
Catalog No. 31501155
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
Standard Taper Joint:
14/20
19/22
24/40
29/42
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Standard Taper Joint
31501155 5 mL 14/20
31501158 25 mL 14/20
31501157 10 mL 19/22
31501156 10 mL 14/20
31501159 25 mL 19/22
31501160 25 mL 24/40
31501161 50 mL 14/20
31501162 50 mL 19/22
31501164 100 mL 14/20
31501163 50 mL 24/40
31501165 100 mL 19/22
31501166 100 mL 24/40
31501167 100 mL 29/42
31501168 200 mL 14/20
31501169 200 mL 24/40
31501170 250 mL 14/20
31501172 250 mL 24/40
31501171 250 mL 19/22
31501173 250 mL 29/42
31501174 500 mL 24/40
31501176 1000 mL 24/40
31501175 500 mL 29/42
31501177 1000 mL 29/42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 31501155 Supplier Synthware™ Supplier No. F311405
Used as an evaporation flask in rotary evaporators
  • WIth standard taper outer joint
  • Heavy wall design for increased safety
  • All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Description Recovery flask
Capacity (Metric) 5 mL
Standard Taper Joint 14/20