Learn More
Synthware™ Reaction Flasks with High Vacuum Valve
GREENER_CHOICE
With high vacuum valve with PTFE O-ring at tip and PTFE wiper to protect Vilton O-ring on shaft
Supplier:  Synthware™ F544500
 View more versions of this product
Catalog No. 31501421
Only null left
Description
Description
SYNTHWARE round bottom single neck reaction flask with a standard taper outer joint and a high vacuum valve sidearm. Useful for sample storage or simple reactions. Valve plug may be removed to sample from the flask while an inert gas purge is passed through the sidearm.
Specifications
Chemical Identifiers
Clear
Borosilicate Glass (3.3)
Organic Synthesis
Reaction flask with high vacuum valve
Specifications
Clear
Borosilicate Glass (3.3)
Reaction flask with high vacuum valve
500 mL
Organic Synthesis
24/40
Product Suggestions
Product Suggestions