missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Reaction Flasks with High Vacuum Valve
GREENER_CHOICE

Catalog No. 31501413
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Standard Taper Joint:
14/20
19/22
24/40
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Standard Taper Joint
31501413 50 mL 14/20
31501414 50 mL 19/22
31501415 100 mL 14/20
31501416 100 mL 19/22
31501417 100 mL 24/40
31501418 250 mL 14/20
31501419 250 mL 19/22
31501420 250 mL 24/40
31501421 500 mL 24/40
31501422 1000 mL 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 31501413 Supplier Synthware™ Supplier No. F541050

With high vacuum valve with PTFE O-ring at tip and PTFE wiper to protect Vilton O-ring on shaft

SYNTHWARE round bottom single neck reaction flask with a standard taper outer joint and a high vacuum valve sidearm. Useful for sample storage or simple reactions. Valve plug may be removed to sample from the flask while an inert gas purge is passed through the sidearm.

Specifications

Color Clear
Capacity (Metric) 50 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Organic Synthesis
Product Type Reaction flask with high vacuum valve
Standard Taper Joint 14/20