missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Reaction Flasks with Glass Stopcock
GREENER_CHOICE

With spring retention device for smooth turning purpose

Supplier:  Synthware™ F534100

 View more versions of this product

Catalog No. 31501432


Only null left

Description

Description

  • Standard taper outer joint
  • Side arm with 2 mm glass stopcock
Specifications

Chemical Identifiers

Reaction flask with glass stopcock

Specifications

100 mL
Glass
Reaction flask with glass stopcock
24/40
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions