Learn More
Synthware™ Reaction Flask, 2-Neck, with Glass Stopcock
With center neck and angled side neck standard taper outer joints of same size
Supplier: Synthware™ F164250
Description
- Side arm with 2 mm glass stopcock with constant pressure retention device
Chemical Identifiers
| Glass |
Specifications
| Joint: 24/40; With center neck and angled side neck standard taper outer joints of same size | |
| 250 mL | |
| Glass | |
| 24/40 |