Learn More
Synthware™ Reaction Flask, 2-Neck, with Glass Stopcock
With center neck and angled side neck standard taper outer joints of same size
Supplier:  Synthware™ F169100
 View more versions of this product
Catalog No. 31501438
Only null left
Description
Description
  • Side arm with 2 mm glass stopcock with constant pressure retention device
Specifications
Chemical Identifiers
Glass
Specifications
Joint: 19/22; With center neck and angled side neck standard taper outer joints of same size
100 mL
Glass
19/22
Product Suggestions
Product Suggestions