Learn More
Synthware™ Reaction Flask, 2-Neck, with Glass Stopcock
GREENER_CHOICE
With center neck and angled side neck standard taper outer joints of same size
Supplier:  Synthware™ KEMF161450
 View more versions of this product
Catalog No. 31501436
Only null left
Description
Description
  • Side arm with 2 mm glass stopcock with constant pressure retention device
Specifications
Chemical Identifiers
Glass
Specifications
Joint: 14/20; With center neck and angled side neck standard taper outer joints of same size
50 mL
Glass
14/20
Product Suggestions
Product Suggestions