Learn More
Synthware™ Powder Funnel
For the introductuion of powders or liquids into reactions
Supplier:  Synthware™ F182450
 View more versions of this product
Catalog No. 31501469
Only null left
Explore available promotions
Description
Description
  • Funnels have 60 deg. side angle and standard taper inner joint
Specifications
Specifications
50mm
Glass
Joint size: 24/40
For the introductuion of powders or liquids into reactions
Product Suggestions
Product Suggestions