Learn More
Synthware™ Kuderna-Danish Flasks
GREENER_CHOICE
Shaped to provide continuous flow of heavier fractions to concentrator tube during reflux
$167.43 - $180.50
Specifications
Type Kuderna-danish Flask
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
31501086
GREENER_CHOICE
View Documents Promotion Details
Synthware™
F984250
250 mL
Each for $167.43
Only null left
31501087
GREENER_CHOICE
View Documents Promotion Details
Synthware™
F984500
500 mL
Each for $180.50
Only null left
Description
Description
  • Top joint: 24/40 outer
  • Lower joint: 19/22 inner
Specifications
Specifications
Kuderna-danish Flask