Learn More
Synthware™ Inlet or Thermometer Adapters
With a compression cap and Viton O-ring to form a vacuum tight seal
$63.77 - $69.17
Specifications
| Material | Borosilicate Glass |
|---|---|
| Product Type | Inlet or Thermometer Adapter |
Description
- Cap accommodates 5.0 to 7.5mm O.D. thermometer
- Supplied with one Nylon™ cap and one Viton™ O-ring
Specifications
| Borosilicate Glass | |
| Inlet or Thermometer Adapter |