Learn More
Synthware™ Inlet Adapters with Stopcock and Drip Tip
With standard taper top outer and lower inner joints
Supplier:  Synthware™ A741922
 View more versions of this product
Catalog No. 31500595
Only null left
Explore available promotions
Description
Description
    • Hose connection used for connection to vacuum source
    • Stopcock allows control of vacuum
    • With 2mm straight-bore PTFE or Glass stopcock
Specifications
Specifications
Stopcock: PTFE
Inlet adapter with stopcock and drip tip
19/22
Product Suggestions
Product Suggestions