missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Inlet Adapters with Hose Connection and Drip Tip
With standard taper top outer and lower inner joints
$106.14 - $140.79
Specifications
| Material | Borosilicate Glass (3.3) |
|---|---|
| Product Type | Inlet adapter with hose connection and drip tip |
| Catalog Number | Mfr. No. | Standard Taper Joint No. | Description | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Standard Taper Joint No. | Description | Price | Quantity | ||||
|
31500591
|
Synthware™
A431420 |
14/20 | Hose O.D.: 8mm |
Each for $106.14
|
|
||||
|
31500592
|
Synthware™
A431922 |
19/22 | Hose O.D.: 8mm |
Each for $108.43
|
|
||||
|
31500593
|
Synthware™
A432440 |
24/40 | Hose O.D.: 10mm |
Each for $140.79
|
|
||||
Description
Hose connection used for connection to a vacuum sourceSpecifications
| Borosilicate Glass (3.3) | |
| Inlet adapter with hose connection and drip tip |