Learn More
Synthware™ Flow Control Adapters with Metering Stopcock, 90°
Allows delivery of preset volumes of gases or liquids to reactions using the needle valve
Supplier:  Synthware™ A721922
 View more versions of this product
Catalog No. 31500582
Only null left
Explore available promotions
Description
Description
Provides on/off control with the main valve
Specifications
Specifications
Borosilicate Glass (3.3)
Flow control adapter
19/22
Product Suggestions
Product Suggestions