Learn More
Synthware™ Flasks with High Vacuum Valve
With high vacuum valve with PTFE O-ring at tip and PTFE wiper (umbrella) to protect Vilton O-ring on shaft
Supplier: Synthware™ F490250
Description
SYNTHWARE storage flask with high vacuum valve and side arm. Valve has a PTFE O-ring at the tip and PTFE wiper to protect the Viton O-ring on the shaft.Specifications
| Clear | |
| 250 mL | |
| Organic Synthesis |
| Valve Size: 0-4mm | |
| Borosilicate Glass (3.3) |