Learn More
Synthware™ Erlenmeyer Flasks, Threaded
For microscale applications
Supplier: Synthware™ F501005
Description
- With 14/20 threaded joint
- Supplied with one holed Nylon compression cap, one Viton O-ring and one PTFE-faced silicone rubber septum
Specifications
| Erlenmeyer Flask | |
| 5 mL | |
| 14/10 |