Learn More
Synthware™ Erlenmeyer Flasks with Outer Joint
With flat bottom and narrow neck
Supplier: Synthware™ F661405
Description
- With standard taper outer joint
- Hand blown from tubing to ensure heavy and uniform wall
Specifications
| Erlenmeyer Flask | |
| 5 mL | |
| 14/20 |