Learn More
Synthware™ Distillation Connecting Adapters, Angled at 75°
For use as a component in atmospheric or vacuum distillations
Supplier: Synthware™ A351014
Description
- Standard taper outer joint at the top
- Inner joints on bottom and side arm are at a 75° angle to one another
Specifications
| Borosilicate Glass (3.3) | |
| Distillation Connecting Adapter | |
| 10/18 (Top), 14/20 (Bottom), 14/20 (Side) |