Learn More
Synthware™ Distillation Adapter, 75°
GREENER_CHOICE
Catalog No. 31500692
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31500692 19/22
31500691 14/20
31500693 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31500692 Supplier Synthware™ Supplier No. A151922
Only null left
Distillation adapter with standard taper inner joints on each end at a 75 deg. angle
Synthware distillation adapter bent at 75 degrees. Both ends are standard taper inner joints and are the same size
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Glass Joint Adapter
Quantity 1
Standard Taper Joint No. 19/22