Learn More
Synthware™ Dewar Flask, Low Form, Hemispherical
Maximum capacity flask fits inside the Dewar.
$896.71 - $1558.75
Specifications
Base Material Borosilicate Glass (3.3)
Material Borosilicate Glass (3.3)
Style Low Form
For Use With (Application) Liquid Nitrogen
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Depth (Metric) Inside Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Depth (Metric) Inside Diameter (Metric) Inner Price Quantity  
31500824
View Documents
Synthware™
F240100
100 mL Flasks 65 mm 80 mm
Each for $896.71
Only null left
31500825
View Documents
Synthware™
F240250
250 mL Flasks 75 mm 100 mm
Each for $1,141.89
Only null left
31500826
View Documents
Synthware™
F240500
500 mL Flasks 85 mm 123 mm
Each for $1,268.00
Only null left
31500827
View Documents
Synthware™
F241000
1000 mL Flasks 100 mm 143 mm
Each for $1,558.75
Only null left
Description
Description
Maximum capacity flask fits inside the Dewar
  • With aluminum housing to transfer magnetic field.
Specifications
Specifications
Borosilicate Glass (3.3)
Borosilicate Glass (3.3)
Low Form
Liquid Nitrogen