Learn More
Synthware™ Dewar Flask, Wide Mouth, Metal Housing
Wide Mouth Dewar Flask with weighted metal base for stability.
Supplier:  Synthware™ F114300
 View more versions of this product
Catalog No. 31500833
Only null left
Explore available promotions
Description
Description
  • Full-length aluminum housing extending to top rim for added protection.
Specifications
Specifications
143 mm
Cylindrical, Wide Mouth
365 mm
305 mm
14.4 in.
4300 mL
7 in
Liquid Nitrogen
Borosilicate Glass (3.3)
178 mm
Borosilicate Glass (3.3)
Product Suggestions
Product Suggestions