Learn More
Synthware™ Dewar Flask, Wide Mouth, Metal Housing
Wide Mouth Dewar Flask with weighted metal base for stability.
Supplier:  Synthware™ F111900
 View more versions of this product
Catalog No. 31500831
Only null left
Explore available promotions
Description
Description
  • Full-length aluminum housing extending to top rim for added protection.
Specifications
Specifications
120 mm
Cylindrical, Wide Mouth
252 mm
190 mm
9.9 in.
1900 mL
6.3 in
Liquid Nitrogen
Borosilicate Glass (3.3)
160 mm
Borosilicate Glass (3.3)
Product Suggestions
Product Suggestions