Learn More
Synthware™ Coil Reflux Condenser, Microscale
GREENER_CHOICE
Catalog No. 31502100
Change view
Click to view available options
Standard Taper Joint No:
14/10
Catalog No. Standard Taper Joint No
31502100 14/10
1 options
Catalog No. 31502100 Supplier Synthware™ Supplier No. C321410
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For low boiling point solvents
  • Supplied with one holed Nylon™ compression cap and one Viton™ O-ring
Specifications
Type Reflux Condenser
Material Glass
Standard Taper Joint No 14/10