Learn More
Synthware™ Chromatography Columns with Standard Taper Joint, PTFE Stopcock and Fritted Disc
Comes in variety of formulation to provide maximum flexibility, control, convenience, and improved yield
Supplier: Synthware™ C184602C
Description
- WIth standard taper top outer joint
- Heavy wall design for increased safety
Specifications
| 53 mm | |
| Chromatography Column | |
| 254 mm |
| 10 in. | |
| 4 mm bore | |
| 60 mm |