Learn More
Synthware™ Chromatography Columns with Standard Taper Joint, PTFE Stopcock and Fritted Disc
Comes in variety of formulation to provide maximum flexibility, control, convenience, and improved yield
Supplier: Synthware™ C184261C
Description
- WIth standard taper top outer joint
- Heavy wall design for increased safety
Specifications
| 20 mm | |
| Chromatography Column | |
| 203 mm |
| 8 in. | |
| 2 mm bore | |
| 26 mm |