Learn More
Synthware™ Chromatography Column with Standard Taper Joint, Reservoir and PTFE Stopcock
For flash chromatography
Supplier:  Synthware™ C378020
 View more versions of this product
Catalog No. 31500898
Only null left
Explore available promotions
Description
Description
  • General purpose chromatography column with reservoir, PTFE stopcock and top standard taper outer joint
  • Heavy-wall design increases safety
Specifications
Specifications
73 mm
24/40
4 mm bore
203 mm
Product Suggestions
Product Suggestions