Learn More
Synthware™ Chromatography Column with Standard Taper Joint, Reservoir and PTFE Stopcock
For flash chromatography
Supplier: Synthware™ C374625
Description
- General purpose chromatography column with reservoir, PTFE stopcock and top standard taper outer joint
- Heavy-wall design increases safety
Specifications
| 40 mm | |
| 24/40 | |
| 2 mm bore | |
| 254 mm |